Learn More
Specifications
Specifications
| Assay Range | 0.78-50 ng/mL |
| Assay Sensitivity | 0.48 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plasma, Serum |
| For Use With (Application) | ELISA, Competitive ELISA |
| Quantity | 1 Kit |
| Research Discipline | Cell Biology, Growth and Development |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Porcine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
