Learn More
Specifications
Specifications
| Assay Range | 15.63-1000 pg/mL |
| Assay Sensitivity | 4.96 pg/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Serum, Plasma |
| Specificity | This kit recognizes Porcine Estradiol in samples. No significant cross-reactivity or interference between Porcine Estradiol and analogues was observed. |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | 17-Beta-Estradiol;17-beta-HSD |
| Target | Porcine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
