Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Porcine IFN-alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. 271051
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
271051 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 271051 Supplier R&D Systems Supplier No. 271051
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 8 publications

IFN-alpha Monoclonal specifically detects IFN-alpha in Porcine samples. It is validated for Neutralization.

Specifications

Antigen IFN-alpha
Applications Neutralization
Classification Monoclonal
Clone F17
Conjugate Unconjugated
Dilution Neutralization
Formulation Supplied as a liquid with No Preservative
Gene Alias IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D
Host Species Mouse
Immunogen Porcine IFN-alpha
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3440
Test Specificity Neutralizes pig interferon alpha
Target Species Pig
Content And Storage After receipt, this product should be kept at -20degreesC for retention of full activity. Do not store below -20degreesC.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.