Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IFN-gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $502.50
Specifications
Antigen | IFN-gamma |
---|---|
Clone | 154007 |
Concentration | LYOPH |
Dilution | Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Porcine samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
IFN-gamma | |
LYOPH | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Pig | |
P17803 | |
3458 | |
E. coli-derived recombinant porcine IFN-gamma Ser21-Lys166 Accession # P17803 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects porcine IFN-gamma in direct ELISAs. |
154007 | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
IFNG | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title