Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IFN-gamma Antibody, R&D Systems™

Goat Antibody
Supplier: R&D Systems AF9851100
This item is not returnable.
View return policy
Description
IFN-gamma specifically detects IFN-gamma in Porcine samples. It is validated for Neutralization.Specifications
IFN-gamma | |
Polyclonal | |
Neutralization 1-6 ng/mL | |
P17803 | |
Goat | |
Antigen Affinity-purified | |
Primary | |
Detects porcine IFN-gamma in direct ELISA. | |
Pig | |
Lyophilized |
Neutralization | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
E. coli-derived recombinant porcine IFN-gamma , Ser21-Lys166, Accession # P17803 | |
100 μg | |
3458 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction