Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IFN-gamma Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF985
This item is not returnable.
View return policy
Description
IFN-gamma Polyclonal specifically detects IFN-gamma in Porcine samples. It is validated for Western Blot, Immunocytochemistry.Specifications
IFN-gamma | |
Polyclonal | |
Biotin | |
P17803 | |
IFNG | |
E. coli-derived recombinant porcine IFN-gamma Ser21-Lys166 Accession # P17803 | |
50 μg | |
Primary | |
Detects porcine IFN-gamma in Western blots. In Western blots, approximately 60% cross-reactivity with recombinant canine IFN-gamma and recombinant feline IFN-gamma is observed, 10% cross-reactivity with recombinant equine IFN-gamma and recombinant human IFN-gamma is observed, and less than 5% cross-reactivity with recombinant mouse IFN-gamma and recombinant rat IFN-gamma is observed. | |
Pig | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
Goat | |
Affinity Purified | |
RUO | |
3458 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction