Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Porcine IgE ELISA Kit (Colorimetric)
SDP

Catalog No. NBP275006 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275006 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275006 Supplier Novus Biologicals™ Supplier No. NBP275006

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Porcine IgE ELISA Kit (Colorimetric) measures porcine IgE in serum, plasma and other biological fluids.

Specifications

Assay Range 0.16 - 10 ng/mL
Assay Sensitivity 0.10 ng/mL
Cross Reactivity This kit recognizes Porcine IgE in samples. No significant cross-reactivity or interference between Porcine IgE and analogues was observed.
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 3497
Gene Symbol IGHE
Host Species Porcine
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target IgE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.