Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Porcine IL-1 beta/IL-1F2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF681
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF681 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF681 Supplier R&D Systems Supplier No. AF681
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

IL-1 beta/IL-1F2 Polyclonal specifically detects IL-1 beta/IL-1F2 in Porcine samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen IL-1 beta/IL-1F2
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 4-20 ug/mL
Gene Accession No. P26889
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Gene Symbols IL1B
Host Species Goat
Immunogen E. coli-derived recombinant porcine IL-1 beta/IL-1F2 Ala115-Pro267 Accession # P26889
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3553
Test Specificity Detects porcine IL-1 beta/IL-1F2 in direct ELISAs and Western blots. In direct ELISAs, approximately 45%-75% cross-reactivity with recombinant human IL-1 beta , recombinant mouse IL-1 beta , recombinant rat IL-1 beta , recombinant feline IL-1 beta , recombinant canine IL-1 beta , recombinant equine IL-1 beta , recombinant guinea pig IL-1 beta , recombinant rabbit IL-1 beta , and recombinant rhesus monkey IL-1 beta is observed and less than 15% cross-reactivity with recombinant cotton rat IL-1 beta is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Pig
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.