Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Porcine IL-10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6931
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6931 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6931 Supplier R&D Systems Supplier No. MAB6931
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

IL-10 Monoclonal specifically detects IL-10 in Porcine samples. It is validated for Neutralization.

Specifications

Antigen IL-10
Applications Neutralization
Classification Monoclonal
Clone 148801
Conjugate Unconjugated
Dilution Neutralization 0.05-0.15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q29055
Gene Alias CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Gene Symbols IL10
Host Species Mouse
Immunogen E. coli-derived recombinant porcine IL-10 Ser19-Asn175 Accession # Q29055
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3586
Test Specificity Detects porcine IL-10 in direct ELISAs. In direct ELISAs, 25-50% cross-reactivity with recombinant feline IL-10, recombinant canine IL-10, and recombinant guinea pig IL-10 is observed, approximately 10% cross-reactivity with recombinant human IL-10 is observed, less than 5% cross-reactivity with recombinant cotton rat IL-10 is observed, and no cross-reactivity with recombinant equine IL-10, recombinant mouse IL-10, recombinant rat IL-10, or recombinant viral IL-10 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Pig
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.