Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IL-10 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF693
This item is not returnable.
View return policy
Description
IL-10 Polyclonal specifically detects IL-10 in Porcine samples. It is validated for Western Blot.Specifications
IL-10 | |
Polyclonal | |
Biotin | |
Q29055 | |
IL10 | |
E. coli-derived recombinant porcine IL-10 Ser19-Asn175 Accession # Q29055 | |
50 μg | |
Primary | |
Detects porcine IL-10 in Western blots. | |
Pig | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
Goat | |
Affinity Purified | |
RUO | |
3586 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction