Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IL-12/IL-35 p35 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC1171G100UG
This item is not returnable.
View return policy
Description
IL-12/IL-35 p35 Monoclonal specifically detects IL-12/IL-35 p35 in Porcine samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-12/IL-35 p35 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q28938, Q29053 | |
Chinese hamster ovary cell line CHO-derived recombinant porcine IL-12 p35/p40 heterodimer p40 Ile23-Asn324, p35 Arg26-Ser222 Accession # Q28938, Q29053 | |
RUO | |
Detects the heterodimeric form of porcine IL-12 (p35/p40) in direct ELISAs. Western blotting data indicates recognition of the IL-12/IL-35 p35 subunit. In direct ELISAs and Western blots, 20-100% cross-reactivity with the disulfide-linked heterodomeric forms of recombinant mouse IL-12 and recombinant human IL-12 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
116219 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Mouse | |
100 μg | |
Primary | |
Pig | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction