Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Porcine IL-18 ELISA Kit

Catalog No. ES12RB
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

ELISA

Porcine IL-18 quantitates porcine IL-18 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant porcine IL-18.

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1 g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320
Assay Range 80 to 20,000 pg/mL
Assay Sensitivity 80 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 397057
Gene Symbol IL18
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IL-18
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.