Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IL-1ra/IL-1F3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | IL-1ra/IL-1F3/IL1RN |
---|---|
Clone | 114801 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Porcine samples. It is validated for Western Blot.Specifications
IL-1ra/IL-1F3/IL1RN | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DIRA, ICIL-1RA, IL1F3, IL-1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
IL1RN | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
114801 | |
Western Blot | |
Unconjugated | |
Mouse | |
Pig | |
Q29056 | |
3557 | |
E. coli-derived recombinant porcine IL-1ra/IL-1F3 His26-Gln177 Accession # Q29056 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects porcine IL-1ra/IL-1F3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse IL-1ra/IL-1F3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title