Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Porcine IL-1ra/IL-1F3 DuoSet ELISA
SDP

Catalog No. DY780 Shop All Novus Biologicals Products
Change view
Click to view available options
Format:
96-well Microplate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
DY780 96-well Microplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY780 Supplier R&D Systems™ Supplier No. DY780

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For determination of natural and recombinant porcine IL-1 receptor antagonist.

The Porcine IL-1ra/IL-1F3 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies porcine IL-1ra/IL-1F3/IL1RN.

Specifications

Product Type Solid Phase Sandwich ELISA
Quantity 1 Kit (for 15 Plates)
Species Porcine
Binding Target IL-1ra/IL-1F3/IL1RN
Assay Range 93.80 to 6000 pg/mL
Format 96-well Microplate
Gene ID (Entrez) 3557 (Human); 16181 (Mouse); 60582 (Rat); 397499 (Porcine)
Product Line R&D Systems
Sample Type Cell Culture Supernatants, Serum, EDTA Plasma, Heparin Plasma
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Immunology
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.