Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IL-2 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC6521S100UG
This item is not returnable.
View return policy
Description
IL-2 Monoclonal specifically detects IL-2 in Porcine samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| IL-2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P26891 | |
| IL2 | |
| E. coli-derived recombinant porcine IL-2 Ala21-Thr154 Accession # P26891 | |
| RUO | |
| 3558 | |
| Pig | |
| IgG2b |
| Flow Cytometry | |
| 100312 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects porcine IL-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with human, mouse, rat, bovine, canine, equine, feline, or cotton rat IL-2 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction