Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IL-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6521
This item is not returnable.
View return policy
Description
IL-2 Monoclonal specifically detects IL-2 in Porcine samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
IL-2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P26891 | |
IL2 | |
E. coli-derived recombinant porcine IL-2 Ala21-Thr154 Accession # P26891 | |
100 μg | |
Primary | |
Detects porcine IL-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with human, mouse, rat, bovine, canine, equine, feline, or cotton rat IL-2 is observed. | |
Pig | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
100312 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3558 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction