Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IL-6 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $409.50
Specifications
Antigen | IL-6 |
---|---|
Clone | 1014310 |
Applications | ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB6862SP
![]() |
R&D Systems
MAB6862SP |
25 μg |
Each for $200.50
|
|
|||||
MAB6862100
![]() |
R&D Systems
MAB6862100 |
100 μg |
Each for $409.50
|
|
|||||
Description
IL-6 Monoclonal specifically detects IL-6 in Porcine samples. It is validated for ELISA.Specifications
IL-6 | |
ELISA | |
Unconjugated | |
Rat | |
Pig | |
P26893 | |
3569 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
1014310 | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
E. coli-derived recombinant porcine IL-6, Pro29-Met212, Accession # P26893 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title