Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IL-6 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems BAF686
This item is not returnable.
View return policy
Description
IL-6 Polyclonal specifically detects IL-6 in Porcine samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
IL-6 | |
Polyclonal | |
Biotin | |
P26893 | |
IL6 | |
E. coli-derived recombinant porcine IL-6 (R&D Systems, Catalog # 686-PI) Pro29-Met212 Accession # P26893 | |
50 μg | |
Primary | |
Detects porcine IL-6 in ELISAs and Western blots. In sandwich immunoassays, less than 0.25% cross-reactivity with recombinant rat IL-6, recombinant mouse IL-6, and recombinant human IL-6 is observed. | |
Pig | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
Goat | |
Affinity Purified | |
RUO | |
3569 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction