Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Porcine IL-8/CXCL8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5351SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5351SP 25 μg
MAB5351100 100 μg
MAB5351500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB5351SP Supplier R&D Systems Supplier No. MAB5351SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

CXCL8/IL-8 Monoclonal specifically detects CXCL8/IL-8 in Porcine samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CXCL8/IL-8
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 105105
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 1-6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. CAA43461
Gene Alias 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
Gene Symbols IL8
Host Species Mouse
Immunogen E. coli-derived recombinant porcine IL-8/CXCL8 Ala26-Gln104 Accession # CAA43461
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3576
Test Specificity Detects porcine IL-8/CXCL8 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha , 12/SDF-1 beta , rhCXCL13, recombinant mouse CXCL1, 2, 6, 9, 10, 12/SDF-1 alpha , rmCXCL13, recombinant rat (rr) CXCL1, rrCXCL3/CINC2 alpha , or rr3/CINC-2 beta is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Pig
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.