Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Porcine Lipocalin-2/NGAL ELISA Kit (Colorimetric)
SDP

Catalog No. NBP262163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262163 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262163 Supplier Novus Biologicals™ Supplier No. NBP262163
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Porcine Lipocalin-2/NGAL ELISA Kit (Colorimetric) measures porcine Lipocalin-2/NGAL in porcine urine, plasma, serum, tissue extracts, culture supernatants.

Specifications

Assay Range 10-400pg/ml
Assay Sensitivity 1 pg/ml
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Plasma, Porcine Urine, Serum, Tissue Extracts
For Use With (Application) ELISA
Gene ID (Entrez) 3934
Gene Symbol LCN2
Host Species Porcine
Quantity 1 Kit
Storage Requirements Store at 4°C
Target Lipocalin-2/NGAL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.