Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Porcine Thyroxine ELISA Kit (Colorimetric)
SDP

Catalog No. NB601581KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB601581KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB601581KT Supplier Novus Biologicals™ Supplier No. NBP2601581KIT
Add to Cart
Add to Cart

Assay has high sensitivity and specificity for detection of pig T4

The Porcine Thyroxine ELISA Kit (Colorimetric) measures porcine Thyroxine in serum, plasma0.

Specifications

Assay Range 20ng/mL to 320ng/mL
Assay Sensitivity 20ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Specificity For detection of pig T4
Target Species Porcine
For Use With (Application) ELISA
Quantity 1 Kit
Research Discipline Signal Transduction
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym T4
Target Thyroxine
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.