Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine TNF-alpha Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems BAF690
This item is not returnable.
View return policy
Description
TNF-alpha Polyclonal specifically detects TNF-alpha in Porcine samples. It is validated for Western Blot.Specifications
TNF-alpha | |
Polyclonal | |
Biotin | |
P23563 | |
TNF | |
E. coli-derived recombinant porcine TNF-alpha (R&D Systems, Catalog # 690-PT) Arg78-Leu232 Accession # P23563 | |
50 μg | |
Primary | |
Detects porcine TNF-alpha in Western blots. | |
Pig | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
Goat | |
Affinity Purified | |
RUO | |
7124 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction