Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PORCN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566168
Description
Immunogen sequence: AVSFHFSNYFV GFLSEATATL AGAGFTEEKD HLEWDLTVSK PLNVELPRSM VEVVTSWNLP MSYWLNNYVF KNAL Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
PORCN belongs to the evolutionarily conserved porcupine (Porc) gene family. Genes of the Porc family encode endoplasmic reticulum proteins with multiple transmembrane domains. Porcupine proteins are involved in the processing of Wnt (wingless and int homologue) proteins. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.This gene belongs to the evolutionarily conserved porcupine (Porc) gene family. Genes of the Porc family encode endoplasmic reticulum proteins with multiple transmembrane domains. Porcupine proteins are involved in the processing of Wnt (wingless and int homologue) proteins. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
PORCN | |
Polyclonal | |
Unconjugated | |
PORCN | |
2410004O13Rik; AW045557; DHOF; DXHXS7465e; FODH; Mg61; MGC29687; mMg61; Mporc; por; PORC; Porcn; porcupine; porcupine homolog; porcupine homolog (Drosophila); porcupine O-acyltransferase; Ppn; probable protein-cysteine N-palmitoyltransferase porcupine; Protein MG61; protein-cysteine N-palmitoyltransferase porcupine; protein-serine O-palmitoleoyltransferase porcupine; RGD1564947 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64840 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H237 | |
PORCN | |
Recombinant Human PORCN. Recombinant protein control fragment (Product #RP-106853). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction