Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PORCN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588445
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588445 Supplier Invitrogen™ Supplier No. PA588445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLNGDRLLRN KKRKARWLRA YESAVSFHFS NYFVGFLSEA TATLAGAGFT EEKDHLEWDL TVSKPLNVEL PRSMVEVVTS WNLPMSYWLN NYVFKNALRL G; Positive Samples: Mouse kidney, Mouse liver, Rat kidney; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.

PORCN belongs to the evolutionarily conserved porcupine (Porc) gene family. Genes of the Porc family encode endoplasmic reticulum proteins with multiple transmembrane domains. Porcupine proteins are involved in the processing of Wnt (wingless and int homologue) proteins. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.This gene belongs to the evolutionarily conserved porcupine (Porc) gene family. Genes of the Porc family encode endoplasmic reticulum proteins with multiple transmembrane domains. Porcupine proteins are involved in the processing of Wnt (wingless and int homologue) proteins. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PORCN
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PORCN
Gene Accession No. Q9JJJ7
Gene Alias 2410004O13Rik; AW045557; DHOF; DXHXS7465e; FODH; Mg61; MGC29687; mMg61; Mporc; por; PORC; Porcn; porcupine; porcupine homolog; porcupine homolog (Drosophila); porcupine O-acyltransferase; Ppn; probable protein-cysteine N-palmitoyltransferase porcupine; Protein MG61; protein-cysteine N-palmitoyltransferase porcupine; protein-serine O-palmitoleoyltransferase porcupine; RGD1564947
Gene Symbols PORCN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 220-320 of human PORCN (NP_9822991).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317368, 53627
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.