Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PORCN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107218
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107218 Supplier Thermo Scientific Supplier No. PA5107218
Add to Cart
Add to Cart

PORCN Polyclonal Antibody

PORCN Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PORCN
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene PORCN
Gene Accession No. Q9JJJ7
Gene Alias 2410004O13Rik; AW045557; DHOF; DXHXS7465e; FODH; Mg61; MGC29687; mMg61; Mporc; por; PORC; Porcn; porcupine; porcupine homolog; porcupine homolog (Drosophila); porcupine O-acyltransferase; Ppn; probable protein-cysteine N-palmitoyltransferase porcupine; Protein MG61; protein-cysteine N-palmitoyltransferase porcupine; protein-serine O-palmitoleoyltransferase porcupine; RGD1564947
Gene Symbols PORCN
Host Species Rabbit
Immunogen Synthetic peptide located within the following region: VAMKAVSLGF DLDRGEVGAV PSPVEFMGYL YFVGTIVFGP WISFHSYLQA.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53627
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.