Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ C1-InhXL Affinity Matrix

Catalog No. 2803402010
Click to view available options
Quantity:
10 mL
50 mL

Thermo Scientific™ POROS™ CaptureSelect™ C1-InhXL Affinity Matrix is developed for the purification of human C1 esterase inhibitor from recombinant and plasma/serum sources.

POROS CaptureSelect C1-InhXL Affinity Matrix is developed for the purification of human C1 esterase inhibitor from recombinant and plasma/serum sources. C1 esterase inhibitor is a protease inhibitor belonging to the serpin superfamily that inhibits the spontaneous activation of the complement system.

POROS CaptureSelect C1-InhXL affinity ligand is highly selective for human C1-inhibitor and coupled to the high-performing POROS matrix, allowing for an efficient and scalable purification process, with high purity and yield obtained in a single step. The large pore structure of the POROS beads enables higher flowrates and efficient binding of larger biomolecules.

Features of POROS CaptureSelect C1-InhXL Affinity Matrix include:

  • Efficient purification of C1 esterase inhibitor with high yield
  • Mild pH elution condition to retain activity
  • High binding capacity
  • Excellent scalability
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: POROS CaptureSelect C1-InhXL affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity:∼3–5 g C1-inhibitor/L resin
  • Elution condition: 50 mM sodium acetate, 1.0 M MgCl2 , pH 5.0
  • Regeneration conditions: acidic stripping condition with 0.5 M acetic acid, pH 2
  • Mechanical resistance: 100 bar (1450 psi, 10 MPa)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Product Line CaptureSelect™
Type Affinity Matrix
Quantity 10 mL
Form Resin Slurry
Particle Size 50 μm
Binding Capacity ∼3 to 5 g/L
Ligand Type C1-InhXL affinity ligand
Matrix Cross-linked poly(styrene-divinylbenzene)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.