Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ CH1-XL Affinity HPLC Columns

Catalog No. A37052
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
100 mm
Diameter (Metric):
2.1 mm
4.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
A37052 30 mm 2.1 mm
A37053 50 mm 4.6 mm
A37054 100 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A37052 Supplier Thermo Scientific™ Supplier No. A37052
Get an Extra Analytical Consumable with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ POROS™ CaptureSelect™ CH1-XL Affinity HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect affinity ligands.

POROS CaptureSelect CH1-XL Affinity HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect affinity ligands.

CaptureSelect CH1-XL Affinity HPLC columns recognize the CH1 domain of human IgG antibodies, enabling quantitation and small-scale purification of all IgG subclasses (IgG1, IgG2, IgG3, and IgG4), as well as Fab fragments and fusion proteins containing the intact CH1 domain, independent of light-chain iso-type and source material.Analytical groups supporting biologic drug development require rapid, sensitive, and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle.

Features of POROS CaptureSelect CH1-XL Affinity HPLC columns include:

  • Versatile—applicable for IgG Fab fragments, all human IgG subclasses, and IgG fusion proteins with intact CH1 domain
  • Specific—no cross-binding to bovine IgG
  • Fast—POROS resins enable quantitation or purification in minutes
  • Hands-off operation—HPLC method can be automated for a streamlined operation
  • Robust—designed to deliver high-performance purification over hundreds of chromatography cycles

High speed with high specificity

POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis, including mass spectrometry.

Automation

POROS Affinity HPLC columns are for use with HPLC systems, which allows the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligand enables one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regimen results in strong re-use performance. POROS Affinity HPLC columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

Main characteristics

  • Protein binding capacity: 26.2 mg/mL for human IgG; 8.8 mg/mL human Fab fragment
  • Species specificity: human
  • Column material: PEEK polymer
  • Frit: 2 μm, titanium
  • POROS resin: cross-linked poly(styrene-divinylbenzene)
  • CaptureSelect ligands: single-domain monospecific antibody fragment produced in Saccharomyces cerevisiae

Specifications

Column Format Analytical Column, Biocompatible PEEK
Diameter (Metric) 2.1 mm
Includes Column includes EZ grip fittings and column test certificate.
Length (Metric) 30 mm
Max. Pressure 2400 psi (170 bar)
Particle Size 20 μm
pH 1-10
Pore Size 2000 Å
Quantity 1 Column
Temperature 2-40°C
Stationary Phase IgG Fab affinity ligand, CH1 domain of human IgG antibodies
Product Line POROS CaptureSelect
Binding Capacity 26.2 mg/ml human IgG 8.8 mg/ml human Fab fragment
Shipping Condition Wet Ice
Packing Material Highly Crosslinked Styrene-Divinylbenzene polymer (ST-DVB)
Column Type Affinity
Phase Affinity Separation
Material PEEK
Technique Bio-LC
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.