Learn More
Thermo Scientific™ POROS™ CaptureSelect™ FibXL Affinity Matrix
Description
POROS CaptureSelect FibXL Affinity Matrix has specifically been designed for the purification of human fibrinogen from recombinant or plasma source.
Features of this affinity matrix include:
- Fibrinogen purification from human plasma and recombinant sources
- Efficient purification of fibrinogen with high yields and retained activity
- Excellent scalability
- Non-animal derived
- High flow rates
POROS CaptureSelect FibXL Affinity Matrix is an affinity matrix that binds to fibrinogen with high selectivity and yield, enabling a robust, fast, and efficient purification process with excellent purity obtained in one step.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics
- Matrix: cross-linked poly(styrene-divinylbenzene)
- Average particle size: 50 μm
- Ligand: POROS CaptureSelect FibXL affinity ligand
- Ligand coupling method: epoxide
- Binding capacity: approximately 15 g/L resin (determined with plasma derived fibrinogen)
- Elution conditions: acidic elution with 0.1 M glycine pH 3.0; neutral pH elution with 20 mM Tris, 1.0 M MgCl2 and 40% (v/v) propyleneglycol
- Mechanical resistance: 100 bar (1450 psi, 10 MPa)
- Formulation buffer: 20 % (v/v) ethanol
Specifications
Specifications
Binding Capacity | 15 g/L |
Matrix | Cross-linked poly(styrene-divinylbenzene) |
Content And Storage | Store in refrigerator (2°C to 8°C). |
Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
Particle Size | 50 μm |
Form | Liquid Suspension |
Product Line | CaptureSelect™ |
Quantity | 10 mL |
Type | Affinity Matrix |
Ligand Type | FibXL affinity ligand |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.