Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ IgM Affinity Column, 4.6 x 50 mm

Catalog No. 4469164
Click to view available options
Quantity:
1 Column

Analytical groups supporting biologic drug development require rapid, sensitive and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates.

Analytical groups supporting biologic drug development require rapid, sensitive and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle.

POROSCaptureSelect IgM HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect ligands. The combination allows three minute quantitation of IgM antibodies.

Features:

  • Broad utility—POROS A, G and CaptureSelectaffinity ligands address most biological drug classes
  • Rapid—POROS resins offer fast quantitation or purification of immunoglobulins and fusion proteins
  • Hands-off operation—HPLC method can be automated for streamlined operation
  • Robust—deliver high-performance purification over hundreds of chromatography cycles

High Speed with High Specificity

The combination of CaptureSelect IgM affinity ligands when combined with POROS 20 μm resin provides a high flow rate, high performance chromatography solution for the rapid quantitation and small-scale purification of IgM.

POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis including mass spectrometry.

Automation

POROS affinity chromatography columns are suitable for use with HPLC systems, which allows the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligands enable one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple Cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regime results in strong re-use performance. POROS Affinity Chromatography columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

LC Lambda Column Specifications:

  • Protein Binding Capacity: 4 mg/mL
  • Species Specificity: Human, Mouse, Rat
  • Column Material: PEEK™ polymer or stainless steel
  • Frit: 2 μm, titanium
  • POROS Resin: Cross-linked poly(styrenedivinylbenzene)
  • CaptureSelect Ligands™: Single-domain monospecific antibody fragment produced in Saccharomyces cerevisiae

Specifications

Binding Capacity 4.0 mg/mL
Capacity (Metric) 0.8 mL
Flow Rate Up to 5000 cm/hr.
Matrix Polystyrenedivinylbenzene
Pore Size 0.16 micron
Content And Storage Column includes EZ grip fittings and column test certificate

Store at 4°C. DO NOT FREEZE.
Diameter (Metric) 4.6 mm
Elution Volume Max 1.6 mL
Elution Volume Min 800 μL
For Use With (Equipment) HPLC
High-throughput Compatibility Not High-throughput Compatible (Manual)
Max. Operating Pressure 180 bar
Length (Metric) 50 mm
Shipping Condition Wet Ice
Reactivity Human, Mouse, Rat
Sample Type Cell Culture Supernatants
Particle Size 20 μm
Product Line CaptureSelect™, POROS™
Quantity 1 Column
Target IgM Antibodies
Type Column
Stationary Phase CaptureSelect™ IgM
Format Column
Column Type Affinity, Pre-packed
Show More Show Less

For Research Use Only. Not for use in diagnostics procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.