Learn More
Thermo Scientific™ POROS™ Ethyl RoboColumn™ Pre-packed columns
Description
This RoboColumn™ product is pre-packed with POROS Ethyl Hydrophobic Interaction Chromatography (HIC) Resin (Cat. No. A32552) and suitable for impurity removal during large-scale downstream purification of biomolecules, primarily for bind/elute operations
RoboColumn™ products are small chromatography columns that are provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms.
Features of POROS Ethyl HIC Resin include:
- Superior resolution capability while maintaining excellent pressure-flow properties, achieved by combining the high-flow characteristics of a POROS base bead with a 50-micron particle size. Higher flow rates help improve productivity.
- Unique selectivity due to the novel hydrophobic ligand, allowing for purification of a wide range of biomolecules, including therapeutic proteins, antibody fragments, antibody drug conjugates, and enzymes
- High capacity, reducing the process volume significantly for subsequent steps
- A rigid polymeric bead with covalent surface chemistry for easier handling and packing with superior physical and chemical stability, allowing cleaning with various clean-in-place (CIP) solutions such as sodium hydroxide and enabling a more robust downstream purification process
These features of POROS Ethyl HIC Resin are a major improvement to the performance of traditional HIC resins, setting the standard for hydrophobic interaction chromatography.
RoboColumn™ is a registered trademark of ATOLL GmbH.
Specifications
Specifications
Product Type | Pre-packed Column |
Quantity | 200 μL |
Content And Storage | Store at room temperature. |
Shipping Condition | Room Temperature |
Product Line | POROS™ |
Stationary Phase | Novel Ethyl |
Capacity (Metric) | 200 μL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.