Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ GoPure™ Oligo (dT)25 Pre-packed Column

Catalog No. p-7191284
Click to view available options
Volume (Metric):
0.2 mL
1 mL
5 mL

Catalog No. A48607

Supplier: Thermo Scientific™ A48607

Only null left
Add to Cart
Add to Cart

Thermo Scientific™ POROS™ GoPure™ pre-packed columns are for use in the screening of POROS affinity, ion exchange, and hydrophobic interaction chromatography (HIC) resins or for small scale purification.

POROS GoPure pre-packed columns are for use in the screening of POROS affinity, ion exchange, and hydrophobic interaction chromatography (HIC) resins or for small scale purification.

These pre-packed columns are ready to connect and have the following features:

  • Available in several dimensions with nominal bed volumes of 0.2, 1, and 5 mL, providing a uniform screening platform
  • Direct connection to standard chromatography systems
  • Packed with POROS high performance resins that offer high dynamic binding capacity and high resolution over a wide range of flow rates
  • Convenient—ready-to-run columns simplify and speed POROS resin evaluations and enable small scale purification
  • Conserves sample—the 0.2 and 1 mL nominal bed volume minimizes sample requirement for screening multiple resins
  • Simplified scalability—the consistency and range of the POROS GoPure family of pre-packed columns simplifies process scale-up and process modeling

POROS GoPure pre-packed columns have been developed to meet the needs of the process development scientist requiring a convenient and uniform POROS resin-screening platform. The smaller bed volumes reduce sample requirements, and the recommended >3-minute residence time (0.3 mL/min) on the 0.5-cm diameter, 5-cm length column equates to 400 cm/hr on a 20-cm bed height. Detailed specifications may be found in the POROS GoPure operating instructions.

POROS Oligo (dT)25 Affinity Resin (Cat. No. A47384) is designed for the purification and isolation of mRNA from in vitro transcription (IVT) manufacturing processes. The resin effectively separates mRNA from components of the transcription reaction process, such as enzymes and plasmid DNA.

Features of POROS Oligo (dT)25 Affinity Resin include:

  • Easy mRNA purification from crude transcription mix to effectively remove plasmid DNA and other IVT components
  • Simplified workflow helps to maximize efficiency, thereby reducing complexity of subsequent polish steps
  • Excellent scalability
  • Non-animal derived

POROS Oligo (dT)25 Affinity Resin allows for a robust platform and efficient purification process for a range of mRNA molecules and applications.

Bulk resin is also offered and comes with a regulatory support package. Worldwide technical support is also available.

Other POROS ion exchange resins, as well HIC and affinity resins, are also available in this column format. All POROS high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates

Specifications

Column Format Pre-packed
For Use With Chromatography Systems
Particle Size 50 μm
Pore Size 200 nm
Quantity 0.2 mL
Stationary Phase Poly deoxythymidine 25 mer (dT-25 mer)
Content And Storage Shipped in 18% ethanol. Store at 2°C to 8°C.
Product Line POROS GoPure
Volume (Metric) 0.2 mL
Column Type Process Development
Purification Target mRNA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.