Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ R2 Prepacked Reversed-Phase Columns

Catalog No. 1111226
Change view
Click to view available options
Particle Size:
10 μm
20 μm
Material:
PEEK
Stainless Steel
Dimensions:
2.1 x 100 mm
2.1 x 30 mm
4.6 x 100 mm
4.6 x 50 mm
Quantity:
1 Column
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Material Dimensions Quantity
11-112-26 10 μm PEEK 4.6 x 100 mm 1 Column
11-112-12 10 μm PEEK 2.1 x 30 mm 1 Column
11-112-16 10 μm PEEK 2.1 x 100 mm 1 Column
11-112-24 10 μm PEEK 4.6 x 50 mm 1 Column
11-114-12 10 μm Stainless Steel 2.1 x 30 mm 1 Column
11-114-16 10 μm Stainless Steel 2.1 x 100 mm 1 Column
11-114-24 10 μm Stainless Steel 4.6 x 50 mm 1 Column
11-114-26 10 μm Stainless Steel 4.6 x 100 mm 1 Column
11-122-12 20 μm PEEK 2.1 x 30 mm 1 Column
11-122-26 20 μm PEEK 4.6 x 100 mm 1 Column
11-124-12 20 μm Stainless Steel 2.1 x 30 mm 1 Column
11-124-26 20 μm Stainless Steel 4.6 x 100 mm 1 Column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 11-112-26 Supplier Thermo Scientific™ Supplier No. 1111226
Only null left
Add to Cart
Add to Cart

Thermo Scientific™ POROS™ Prepacked Reversed-Phase Column. POROS R2 is a 2000 angstrom pore size Poly[styrenevinylbenzene] particle, suitable for the reversed-phase separation of biomolecules.

POROS R2 is a 2000 angstrom pore size Poly[styrenevinylbenzene] particle, suitable for the reversed-phase separation of biomolecules. 10 micron particle size is used for the highest resolution chromatography, and is suitable for analytical to small scale preparative separation of biomolecules. 20 micron particle size is used for high resolution and small scale preparative to semi-preparative separation of biomolecules.

POROS High Performance Chromatography™ Media and Pre-Packed Columns

  • A high performance chromatography resins for analytical to process scale separations.
  • Higher productivity: High throughput and high dynamic capacity associated with High Performance Chromatography™
  • Chemical stability: Allows aggressive cleaning and sanitization
  • Enhanced biomolecule access: Provided via large pores, ranging between 500-10,000 Å
  • Polystyrenedivinylbenzene particles: Yield a robust, easily packable matrix
  • Develop better separations methods in a shorter time frame: The speed of High Performance Chromatography technology reduces weeks of experimentation to only a few hours of work, so you have plenty of time to explore all the variables of your separation

Reduce time-consuming sample prep

You can replace many sample preparation steps with high speed High Performance Chromatography technology. For instance, dialysis of large volumes of material can be replaced by high flow rate processing of the dilute sample. Elution in a small volume of new buffer accomplishes both buffer exchange and concentration.

Create novel assays for more efficient analysis

High Performance Chromatography technology is not limited to standard modes of separation. Both enzymes and affinity ligands can be immobilized on POROS media. By combining rapid on-column protein digestions with rapid on-column immunoassays and chromatographic separations, you can create entirely new assays with unlimited potential.

High capacity, high resolution, high speed

In contrast to conventional chromatography media, POROS High Performance Chromatography™ resins particles, are engineered to have two discreet classes of pores. Large 'throughpores' allow convection flow to occur through the particles themselves, quickly carrying sample molecules to short 'diffusive' pores inside. By reducing the distance over which diffusion needs to occur, the time required for sample molecules to interact with interior binding sites is also reduced. Diffusion is no longer limiting and flow rates can be dramatically increased–without compromising resolution or capacity. Separations can be achieved at 1000 to 5000 cm/hr compared to 50 to 360 cm/hr for conventional media.

Specifications

Column Format Analytical Column, Biocompatible PEEK
Diameter (Metric) 4.6 mm
Length (Metric) 100 mm
Max. Pressure 2300 psi (200 bar)
Particle Size 10 μm
pH 1-14
Pore Size 2000 Å
Quantity 1 Column
Temperature <80°C
Stationary Phase Phenyl
Content And Storage Store at room temperature.
Product Line POROS
Binding Capacity 10 mg/mL (Lysozyme, 1% acetonitrile)
Shipping Condition Room Temperature
Volume (Metric) 1.7 mL
Dimensions 4.6 x 100 mm
Packing Material Highly Crosslinked Styrene-Divinylbenzene polymer (ST-DVB)
Column Type Reversed Phase
Material PEEK
Technique Bio-LC
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.