Learn More
Thermo Scientific™ POROS™ XS Strong Cation Exchange Resin, 50 mL PROMO
Description
- Dynamic binding capacity leads to reduced column size, smaller footprint, decreased buffer usage, and reduced cycling
- High resolution supports better product yield and improved impurity clearance
- Robust salt tolerance allows improved process flexibility, alignment, and process flow streamlining
- Low back pressure and linear pressure responses help improve scalability
- Rigid polymeric bead with covalent surface chemistry makes for easier handling and packing, and superior physical and chemical stability, resulting in a robust manufacturing process
Dynamic binding capacityPOROS XS resin has a high binding capacity of more than 100 mg/mL, at 5% breakthrough, over a broad range of process conditions.Higher dynamic binding capacity was achieved by optimizing the base bead pore structure and surface chemistry.
High resolution
Using an optimized 50 μm particle size, POROS XS resin maintains the superior resolving capacity of POROS™ HS resin while overcoming the challenging impurity levels associated with high titer processes. Resolution is consistent across a wide range of operating flow rates and is maintained independent of protein load.
Robust salt tolerance
POROS XS resin has high and consistent protein capacity across a broad range of salt concentrations up to 150 mM NaCl (15 mS/cm conductivity). The ability to load columns under higher conductivity reduces or helps eliminate the dilution of column feed streams or the removal of unit operations, such as tangential flow filtration. The optimized surface ionic capacity of POROS XS resin minimizes competitive binding between the protein of interest and the counterion, resulting in superior salt tolerance.
Specifications
Specifications
| Quantity | 50 mL |
| Product Line | POROS™ XS Strong Cation Exchange Resin |
| Concentration or Composition (by Analyte or Components) | Crosslinked Poly(styrene-divinylbenzene) |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.