Learn More
Porosity of Soils
SDPSCI_ED
Catalog No. S05974
Click to view available options
:
Soil Porosity Kit
Catalog No. S05974
Supplier: Science First 6217150
Kit shows the relationship between particle size, shape and uniformity.
  • Place two different soil samples in the side-by-side high-quality tubes
  • Using the included syringes, insert an equal amount of water in each
  • Measure the rate of flow to determine porosity
  • Fully assembled unit has precision-cut sytrene housing
Specifications
Description Porosity of Soils Apparatus
Includes High-quality tubes, syringes, precision-cut sytrene housing
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.