Learn More
Eisco™ Porphyritic Granite Specimen
SCI_ED
Catalog No. p-7076850 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24758 1 Ea. 2 to 3 cm
S26612 10/Pk. 2 to 3 cm
S24759 12/Pk. 2 to 3 cm
S26658 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24758 Supplier Eisco™ Supplier No. ESNG0044
Only null left
Add to Cart
Add to Cart
Porphyritic Granite Specimen is a must-have sample when discussing rock types, rock cycle, formation, classification, and identification of igneous rocks.
  • Large crystals in variety of shapes and colors
  • Felsic plutonic rock ideal for lessons in mineral identification and bulk rock chemistry
  • For comparing cooling rates between samples with differently-sized crystals
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Large crystals are indicative of slow cooling rate, and make mineral identification simple
Specifications
Product Type Igneous Rock, Porphyritic Granite
For Use With (Application) Comparing cooling rates between samples with differently-sized crystals
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm