Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDTOX™ Positive Control

Catalog No. p-7074250
Click to view available options
Intended Use:
Positive control used for quality control with MEDTOX PROFILE-II ER and all VERDICT -II products
Positive control used for quality control with all MEDTOX PROFILE-II tests except ER
Type:
MEDTOX 2000 Positive Toxicology Control
MEDTOX 300 Positive Toxicology Control

Catalog No. 22-000-402

Supplier: MEDTOX™ 102366

Due to product restrictions, please to purchase or view availability for this product.

Designed to monitor the performance of MEDTOX™ drugs of abuse test devices. MEDTOX™ Positive Controls are compatible with all quantitative and qualitative drug detection procedures, which are sufficiently sensitive to detect the control constituents.

  • MEDTOX 300 Positive Toxicology Control: Use for Opiate 300 tests (PROFILE-II ER and all VERDICT -II products)
  • MEDTOX 2000 Positive Toxicology Control: Use for Opiate 2000 tests (All PROFILE-II except ER)
  • Positive control urines have been spiked with authentic reference drug standards and/or appropriate metabolites
  • Should be treated as any “unknown” specimen while following the specific protocol of the assay being used
  • Controls prepared from human based urine available as a negative and Cutoff +50% to monitor the performance of MEDTOX drugs of abuse test devices
  • Each bottle contains stabilized human based urine
Storage Information
Unopened:

The controls are stable for two years or until the expiration date, whichever comes first, when stored at -20°C protected from light.

The controls are stable for six months or until the expiration date, whichever comes first, when stored at 2 to 8°C

After Opening:

The controls are stable for six months or until the expiration date, whichever comes first, when stored at -20°C (Controls can be aliquoted and frozen)

The controls are stable for one month or until the expiration date, whichever comes first, when stored at 2 to 8°C

Specifications

Sample Type Urine
Type MEDTOX 2000 Positive Toxicology Control
Intended Use Positive control used for quality control with all MEDTOX PROFILE-II tests except ER
Packaging Bottle
Quantity 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.