Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POT1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500176MFV450
Description
POT1 Polyclonal antibody specifically detects POT1 in Human samples. It is validated for Western BlotSpecifications
POT1 | |
Polyclonal | |
Western Blot | |
DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1 | |
A synthetic peptide made to an internal portion of the human POT1 protein sequence (between residues 200-300). [UniProt# Q9NUX5] | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing | |
25913 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction