Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APPA6830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APPA6830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APPA6830 Supplier Invitrogen™ Supplier No. PA116830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The target sequence has 95% sequence homology with monkey, 82% sequence homology with rat, and 72% sequence homology with mouse. Suggested positive control: HeLa nuclear extract, antigen standard for POT1 (transient overexpression lysate).

POT1 is a component of the telomerase ribonucleoprotein (RNP) complex that is essential for the replication of chromosome termini. It is a component of the double-stranded telomeric DNA-binding TRF1 complex which is involved in the regulation of telomere length by cis-inhibition of telomerase. Pot1 also acts as a single-stranded telomeric DNA-binding protein and thus may act as a downstream effector of the TRF1 complex and may transduce information about telomere maintenance and/or length to the telomere terminus. It binds to two or more telomeric single-stranded 5'-TTAGGG-3' repeats (G-strand) and with high specificity to a minimal telomeric single-stranded 5'-TAGGGTTAG-3' sequence. Its activity is TERT dependent but it does not increase TERT activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POT1
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene POT1
Gene Accession No. Q9NUX5
Gene Alias 1500031H18Rik; AI505244; AI851169; CMM10; DKFZp586D211; Dolichyl-phosphate-mannose--protein mannosyltransferase 1; GLM9; HPOT1; mPot1; Pomt1; POT 1; POT1; POT-1; Pot1a; POT1-like telomere end-binding protein; protection of telomeres 1; protection of telomeres 1 homolog; protection of telomeres 1A; protection of telomeres protein 1; protein O-mannosyltransferase 1; Protein O-mannosyl-transferase 1; protein-O-mannosyltransferase 1
Gene Symbols POT1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human POT1 protein sequence (between residues 200-300).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25913
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.