Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531544
Description
Recommended positive controls: 293T, A431, Jurkat. Predicted reactivity: Pig (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
POT1 is a component of the telomerase ribonucleoprotein (RNP) complex that is essential for the replication of chromosome termini. It is a component of the double-stranded telomeric DNA-binding TRF1 complex which is involved in the regulation of telomere length by cis-inhibition of telomerase. Pot1 also acts as a single-stranded telomeric DNA-binding protein and thus may act as a downstream effector of the TRF1 complex and may transduce information about telomere maintenance and/or length to the telomere terminus. It binds to two or more telomeric single-stranded 5'-TTAGGG-3' repeats (G-strand) and with high specificity to a minimal telomeric single-stranded 5'-TAGGGTTAG-3' sequence. Its activity is TERT dependent but it does not increase TERT activity.
Specifications
| POT1 | |
| Polyclonal | |
| Unconjugated | |
| POT1 | |
| 1500031H18Rik; AI505244; AI851169; CMM10; DKFZp586D211; Dolichyl-phosphate-mannose--protein mannosyltransferase 1; GLM9; HPOT1; mPot1; Pomt1; POT 1; POT1; POT-1; Pot1a; POT1-like telomere end-binding protein; protection of telomeres 1; protection of telomeres 1 homolog; protection of telomeres 1A; protection of telomeres protein 1; protein O-mannosyltransferase 1; Protein O-mannosyl-transferase 1; protein-O-mannosyltransferase 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25913 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.73 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NUX5 | |
| POT1 | |
| Recombinant protein encompassing a sequence within the center region of human POT1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction