Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 105811AP150UL

Description
POT1 is a component of the telomerase ribonucleoprotein (RNP) complex that is essential for the replication of chromosome termini. It is a component of the double-stranded telomeric DNA-binding TRF1 complex which is involved in the regulation of telomere length by cis-inhibition of telomerase. Pot1 also acts as a single-stranded telomeric DNA-binding protein and thus may act as a downstream effector of the TRF1 complex and may transduce information about telomere maintece and/or length to the telomere terminus. It binds to two or more telomeric single-stranded 5′-TTAGGG-3′ repeats (G-strand) and with high specificity to a minimal telomeric single-stranded 5′-TAGGGTTAG-3′ sequence. Its activity is TERT dependent but it does not increase TERT activity.Specifications
POT1 | |
Polyclonal | |
Unconjugated | |
POT1 | |
hPot1, POT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
101185, 25913 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q91WC1, Q9NUX5 | |
Pot1 | |
POT1 Fusion Protein Ag0875 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction