Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potash, sulfurated, S 11.8% min as sulfide

Catalog No. p-4407712
Click to view available options
Quantity:
100 g
500 g

Mixture composed of potassium polysulfides and potassium thiosulfate | CAS: 39365-88-3 | K4O3S3 | 300.57 g/mol

Potash is used in black/brown patina (copper, brass, bronze). Its use include decorative color effects on copper, brass, bronze and nickel. Sulfurated potash has been used in the treatment of acne and other skin disorders usually in the form of a lotion with zinc sulfate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39365-88-3
Molecular Formula K4O3S3
Molecular Weight (g/mol) 300.57
MDL Number MFCD00064604
InChI Key YFKFGPGTVQHNEF-UHFFFAOYSA-L
Synonym potash, sulfurated, sulfurated potash, potash, sulfurated usp, liver of sulfur, 2k.k2s.so32, potash sulfurated liver of sulfur, dipotassium hypo potassium sulfide, potash, sulfurated, s as sulfide, dipotassium thiosulfate mixture with potassium sulfide, dipotassium thiosulfate mixture with potassium sulfide k2sx
PubChem CID 56842985
IUPAC Name tetrapotassium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;sulfanide
SMILES [S--].[K+].[K+].[K+].[K+].[O-]S([O-])(=O)=S

Specifications

Color Green
Physical Form Chunks
Quantity 100 g
Assay Percent Range S 11.8% min as sulfide
Solubility Information Soluble in water
Sensitivity Air sensitive; Hygroscopic
Appearance Green chunks
Chemical Name or Material Potash, Mixture composed of potassium polysulfides and potassium thiosulfate
Hazard Category H228-H314-H335
Precautionary Statement P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.1, Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.