Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium 2-ethylhexanoate, 99.9% (metals basis), 75% w/w soln.

Catalog No. p-7050634
Click to view available options
Quantity:
2 g
50 g

CAS: 3164-85-0 | C8H15KO2 | 182.30 g/mol

Used to convert the tert-butylammmonium salt of clavulanic acid into potassium clavulanate or clavulanate potassium. Also used as a catalyst for polyurethane systems (foams) and for unsaturated polyester resin systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3164-85-0
Molecular Formula C8H15KO2
Molecular Weight (g/mol) 182.30
MDL Number MFCD00045896
InChI Key ZUFQCVZBBNZMKD-UHFFFAOYNA-M
PubChem CID 23669619
SMILES [K+].CCCCC(CC)C([O-])=O

Specifications

Odor Characteristic
Linear Formula K[OOCCH(C2H5)C4H9]
Quantity 2 g
UN Number UN1993
Synonym potassium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, potassium salt, 2-ethylhexanoic acid potassium salt, 2-ethylhexanoic acid, potassium salt, hexanoic acid, 2-ethyl-, potassium salt 1:1, potassium2-ethylhexanoate, dsstox_cid_7525, dsstox_rid_78488, dsstox_gsid_27525, ksc493c3p
Solubility Information Partly miscible with water.
IUPAC Name potassium;2-ethylhexanoate
Formula Weight 182.3
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 75% w/w soln.
Physical Form Viscous Liquid
Assay (metals basis)
Chemical Name or Material Potassium 2-ethylhexanoate
Show More Show Less
Hazard Category H315-H318-H361
Hazard Statement GHS H Statement
H226-H361-H373
Flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P264b-P281-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 221-625-7
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.