Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium 4-methoxyphenyltrifluoroborate, 95%

Catalog No. p-7029870
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2738403 1 g
AAH2738406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2738403 Supplier Thermo Scientific Chemicals Supplier No. H2738403
Only null left
Add to Cart

CAS: 192863-36-8 | C7H7BF3KO | 214.036 g/mol

It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192863-36-8
Molecular Formula C7H7BF3KO
Molecular Weight (g/mol) 214.036
MDL Number MFCD04112717
InChI Key XNYMCUFKZHRYID-UHFFFAOYSA-N
Synonym potassium trifluoro 4-methoxyphenyl borate, potassium 4-methoxyphenyl trifluoroborate, potassium 4-methoxyphenyltrifluoroborate, potassium trifluoro 4-methoxyphenyl boranuide, pubchem11347, amtb103, potassum 4-methoxyphenyl trfluoroborate, trifluoro 4-methoxyphenyl potassioboron v, potassium trifluoro-4-methoxyphenyl boranuide
PubChem CID 23683762
IUPAC Name potassium;trifluoro-(4-methoxyphenyl)boranuide
SMILES [B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+]

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 214.04
Percent Purity 95%
Chemical Name or Material Potassium 4-methoxyphenyltrifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.