Learn More
Potassium Acid Phthalate, 300 mg/L, Multi-Purpose Standard, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Color | Colorless |
| CAS Min % | 98.0, 0.03 |
| CAS Max % | 100.0, 0.03 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 300mg/L, Multi-Purpose Standard |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Potassium Acid Phthalate |
Safety and Handling
| ShelfLife | 12 months |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.