Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Antimony Tartrate Solution, For Phosphorus, LabChem™

Catalog No. lc187202
Click to view available options
Quantity:
500 mL
1 L

Specifications

Chemical Name or Material Potassium Antimony Tartrate Solution
Color Colorless
Name Note For Phosphorus
CAS Max % 99.23, 0.495, 0.27, 0.005
CAS 67-66-3, 28300-74-5, 64-17-5
Linear Formula C8H4K2O12Sb2·3H2O
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 667.87
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Antimony and its oxides; Carbon monoxide; Carbon dioxide
Quantity 1 L
Physical Form Liquid
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.