Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Antimony Tartrate Solution, For Phosphorus, LabChem™
Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Potassium Antimony Tartrate Solution |
Color | Colorless |
Name Note | For Phosphorus |
CAS Max % | 99.23, 0.495, 0.27, 0.005 |
CAS | 67-66-3, 28300-74-5, 64-17-5 |
Linear Formula | C8H4K2O12Sb2·3H2O |
Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
Solubility Information | Soluble in water |
Formula Weight | 667.87 |
Identification | Passes Test |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction