Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium benzoate, 99%

Catalog No. p-7081579
Click to view available options
Quantity:
250 g
1000 g

CAS: 582-25-2 | C7H5KO2 | 160.213 g/mol

Potassium Benzoate is manufactured primarily for food and beverage use. It is a chemical preservative, which in very low concentrations inhibits the activity of the microorganisms. It is used in carbonated beverages. The shelf life of un-pasteurized cider can be greatly extended by adding potassium benzoate. It is also used as the whistle in many fireworks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium Benzoate is manufactured primarily for food and beverage use. It is a chemical preservative, which in very low concentrations inhibits the activity of the microorganisms. It is used in carbonated beverages. The shelf life of un-pasteurized cider can be greatly extended by adding potassium benzoate. It is also used as the whistle in many fireworks.

Solubility
Soluble in ethanol, slightly soluble in methanol, insoluble in ether.

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 582-25-2
Molecular Formula C7H5KO2
Molecular Weight (g/mol) 160.213
MDL Number MFCD00013061
InChI Key XAEFZNCEHLXOMS-UHFFFAOYSA-M
Synonym potassium benzoate, benzoic acid, potassium salt, potassium salt, unii-763yqn2k7k, potassium benzoate nf, benzoic acid potassium salt, potassium ion benzoate, acmc-20ajv6, dsstox_cid_7219
PubChem CID 23661960
IUPAC Name potassium;benzoate
SMILES C1=CC=C(C=C1)C(=O)[O-].[K+]

Specifications

Melting Point >300°C
Assay Percent Range 99%
Quantity 250 g
Beilstein 3719165
Sensitivity Hygroscopic
Merck Index 14,1091
Solubility Information Soluble in ethanol,slightly soluble in methanol,insoluble in ether.
Formula Weight 160.22
Percent Purity 99%
Chemical Name or Material Potassium Benzoate
EINECSNumber 209-481-3
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.