Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bicarbonate (Cryst./Certified ACS), Fisher Chemical

Catalog No. p184500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Melting Point 100°C
Color White
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range 99.7 to 100.5%
Formula Weight 100.12
Percent Purity 99.7 to 100.5%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Decomposition Information >100°C
Density 2.17g/cm³
Chemical Name or Material Potassium Bicarbonate
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May be harmful by inhalation, in contact with skin and if swallowed. Wear personal protective equipment. Use personal protective equipment. Look out! DATATYPE 710. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. . . . .

NFPA

Health:1
Flammability:0
Instability:0
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.