Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bicarbonate, Granular, FCC, 99-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18605454
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

P1182, 298-14-6, KHCO3

Spectrum™ Chemical Potassium Bicarbonate, Granular, FCC is used as a carbon dioxide source for leavening in baking. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99 to 101.5%
Grade FCC
Loss on Drying 0.0025
Packaging Poly Pail
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.