Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Biiodate, 0.0250 N, 0.002083 M, Ricca Chemical
SDP

Catalog No. 587532 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 13455-24-8, 7732-18-5
Molecular Formula HI2KO6
Molecular Weight (g/mol) 389.909
InChI Key ACAYDTMSDROWHW-UHFFFAOYSA-M
Synonym potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
PubChem CID 23700942
IUPAC Name potassium;iodic acid;iodate
SMILES OI(=O)=O.[O-]I(=O)=O.[K+]

Specifications

Color Colorless
CAS Min % 98.0, 0.08
CAS Max % 100.0, 0.08
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.0250Normal,0.002083Molar
Chemical Name or Material Potassium Biiodate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.